Thermo Scientific™ Super-Nuova™ Single-Position Digital Stirrers
Obtain unmatched performance, safety and control with Thermo Scientific™ Super-Nuova™ Single-Position Digital Stirrers.
$1,764.56 - $1,764.56
Specifications
Material Ceramic
Speed 50 to 1,200rpm
Stirring Positions 1
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Voltage Price Quantity  
Catalog Number Mfr. No. Capacity Voltage Price Quantity  
11675924Q Thermo Scientific™
S133325Q
4L 120V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
09047120Q Thermo Scientific™
S13393033Q
6L 220/240V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
09050288Q Thermo Scientific™
S133935Q
6L 120V Each for $1,764.56
Specifications
Description
Obtain unmatched performance, safety and control with Thermo Scientific™ Super-Nuova™ Single-Position Digital Stirrers, which deliver easy operation and sophisticated controls for maximum safety. Thermo Scientific™ StirTrac™ technology offers improved coupling and stronger magnetic control. Ceramic-top general-purpose stirrers with four-function memory are ideal for labs with multiple users.
Operating Features
  • Microprocessor control ensures accurate and stable setpoints for stirring speed
  • Single control knob controls speed; lock feature prevents accidental changes
  • Digital rpm display
  • Seamless, reflective white ceramic surface aids sample visibility
  • Stirring adjustable in units of 1rpm
  • RS-232 port outputs time count, stir speed (setpoint) directly to PC
Safety Features
  • Rugged low-profile cast-aluminum body provides stability and durability
  • Unit design protects internal components from accidental spills
  • Walk-away timer shuts off stirring after 1, 2, 4, 8, or 12 hrs.
Includes:
  • Detachable line cord and plug
Description & Specifications
Specifications
Ceramic
50 to 1,200rpm
1