Super Models
Observe the properties of various compounds.
Manufacturer:  Fisher Science Education BKY60
 View more versions of this product
Catalog No. S65374
Description
Description
The Buckyball model illustrates the stability of the structure, and the ability to bounce and trap atoms within the cage. It includes 62 trigonal carbons and 92 vinyl tubes.
The Diamond model contains 57 tetrahedral carbons and 82 vinyl tubes. It illustrates the stability and hardness of diamond, network covalent bonding, and cleavage.
The Sodium Chloride model exhibits 6:6 coordination and a cubic crystal system. It includes 33 sodium ions, 33 chloride ions, and 144 vinyl tubes.
Specifications
Specifications
Education
19.05cm
62 Trigonal planar Carbon atom centers, 1.25 in. PVC clear bonds, assembly instruction booklet
7.5 in.
Stability of the structure
C60 bulkyball model set
Documents