missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Super Models

Observe the properties of various compounds.

$25.00 - $25.00

Specifications

For Use With (Application) Stability of the structure
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Type Price Quantity  
Catalog Number Mfr. No. Includes Type Price Quantity  
S65374 Fisher Science Education
BKY-60
62 Trigonal planar Carbon atom centers, 1.25 in. PVC clear bonds, assembly instruction booklet C60 bulkyball model set Each for $25.00
Specifications

Description

The Buckyball model illustrates the stability of the structure, and the ability to bounce and trap atoms within the cage. It includes 62 trigonal carbons and 92 vinyl tubes.
The Diamond model contains 57 tetrahedral carbons and 82 vinyl tubes. It illustrates the stability and hardness of diamond, network covalent bonding, and cleavage.
The Sodium Chloride model exhibits 6:6 coordination and a cubic crystal system. It includes 33 sodium ions, 33 chloride ions, and 144 vinyl tubes.

Description & Specifications

Specifications

Stability of the structure