American Educational Products Sunpower House
Teach the principles of passive solar heating.
Manufacturer:  American Educational Products HUB4706
 View more versions of this product
Catalog No. S42212B
Description
Description
A working model of a passive solar house that teaches students concepts such as the Greenhouse Effect, utilization of solar heat energy, advantages of different materials to absorb solar radiation, use of insulation and reflectors, and energy conservation.
Roof of the house may be opened to:
  • Insert the thermometer
  • Change the color of the floor panels
  • Replace the solar mass with water or rock heat storage
Specifications
Specifications
11 x 11 x 8 in. (27.9 x 27.9 x 20.3cm)
Thermometer, Water storage container, dark and light modular floor panels, a combination insulation/reflector panel, teachers guide
11 in.
8 in.
11 in.
3 to 9
Solar Energy Apparatus
27.9cm
20.3cm
27.9cm
Documents