Please login to your online account to display your discounted pricing

Sunitinib malate, Tocris™

Potent VEGFR, PDGFRβ and KIT inhibitor

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material Sunitinib malate
Assay Percent Range >99%
Molecular Formula C22H27FN4O2·C4H6O5
Formula Weight 532.56
Synonym N-[2-(Diethylamino)ethyl]-5-[(Z)-(5-fluoro-1, 2-dihydro-2-oxo-3H-indol-3-ylidine)methyl]-2, 4-dimethyl-1H-pyrrole-3-carboxamide (2S)-2-hydroxybutanedioate salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

376850

 
r & d systems
RDS376850
50mg Each for $1,116.26

376810

 
r & d systems
RDS376810
10mg Each for $265.15
Description & Specifications

Specifications

Formula Weight 532.56
Quantity 50mg
Synonym N-[2-(Diethylamino)ethyl]-5-[(Z)-(5-fluoro-1, 2-dihydro-2-oxo-3H-indol-3-ylidine)methyl]-2, 4-dimethyl-1H-pyrrole-3-carboxamide (2S)-2-hydroxybutanedioate salt
Chemical Name or Material Sunitinib malate
Assay Percent Range >99%
CAS 341031-54-7
Molecular Formula C22H27FN4O2·C4H6O5
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C