missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SUN-SRi™ StepVial System™ Vials

2mL, 12 × 32mm Crimp/Snap Ring Vials

Manufacturer:  Thermo Scientific™ 501302

 View more versions of this product

Catalog No. 14-823-372



Description

Description

  • Wide-opening, expansion, borosilicate glass vials
  • Fit rotating or robotic arm autosamplers
  • I-D™ vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • System design automatically seats inserts at optimal center and position for injection

Specifications

Specifications

11mm
1.25 in.
Yes
100 Pack
StepVial™ Seal Vial
Amber
Borosilicate Glass
Crimp
32mm
Wide
Flat
<170μL
12 mm
2.0 mL
Documents