Thermo Scientific™ SUN-SRi™ Filter Tips
Economical dissolution filters
Manufacturer:  Thermo Scientific™ 204004
 View more versions of this product
Catalog No. 14-823-236
Description
Description
  • Polyethylene with a limited porous area
  • Fit securely on the end of a 1/8 in. cannula or sample probe
Specifications
Specifications
1/8 in. Cannula or Sample Probe
1000/Cs.
Dissolution Filter Tip
10 µm
Polyethylene