Thermo Scientific™ SUN-SRi™ 20mm Crimp-Top and 18mm Screw-Top Headspace Vials: Round Bottom
Designed with a round bottom that is more easily handled by robotic arms
$81.55 - $146.25
Specifications
Material Glass
Type Headspace Vial
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Closure Type Includes Color Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Type Includes Color Price Quantity  
36-100-5029 Thermo Scientific™
502046
10.0mL Crimp Clear Pack of 125 Each for $97.47
36-100-5030 Accessories Thermo Scientific™
502458
20.0mL Screw cap Clear Pack of 125 Each for $120.68
36-100-5031 Thermo Scientific™
502460
10.0mL Screw cap Cap Not Included Clear Pack of 125 Each for $84.85
14-823-340 Thermo Scientific™
500550
20.0mL Crimp Clear Pack of 100 Each for $81.55
14-823-531 Thermo Scientific™
502554
20.0mL Crimp Amber Pack of 125 Each for $146.25
14-823-450 Thermo Scientific™
502459
20.0mL Screw cap Cap Not Included Clear Pack of 100 Each for $83.65
Specifications
Description
  • Available with a 20mm crimp-seal or 18mm screw-thread finish; caps sold separately
  • Compatible with Gerstel™ and CTC/Leap™ Technologies headspace samplers when used with magnetic closures
  • Beveled-edge finish on crimp-top vials provide for a sturdy rim that presses into the septum for a more effective seal
Description & Specifications
Specifications
Glass
Headspace Vial