missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SUN-SRi™ 20mm Crimp-Top and 18mm Screw-Top Headspace Vials: Round Bottom

Designed with a round bottom that is more easily handled by robotic arms.

Manufacturer:  Thermo Scientific™ 502460

 View more versions of this product

Catalog No. 36-100-5031



Description

Description

  • Available with a 20mm crimp-seal or 18mm screw-thread finish; caps sold separately
  • Compatible with Gerstel™ and CTC/Leap™ Technologies headspace samplers when used with magnetic closures
  • Beveled-edge finish on crimp-top vials provide for a sturdy rim that presses into the septum for a more effective seal

Specifications

Specifications

Screw cap
18mm
22 mm
Headspace Vial
1.77 in.
Cap Not Included
Round
Tin Plated Metal
Clear
Glass
10 mL
45mm
125 Pack
Documents