missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Sumatriptan succinate

5-HT1A/1B/1D receptor agonist

$112.75 - $465.94

Specifications

Product Type Sumatriptan succinate
Inhibitors Sumatriptan succinate
Molecular Weight 413.49
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
358610R Tocris™
RDS358610
10mg Each for $112.75
358650R Tocris™
RDS358650
50mg Each for $465.94
Specifications

Description

5-HT1 receptor agonist (Ki values are 17, 27 and 100nM at 5-HT1D, 5-HT1B and 5-HT1A receptors respectively). Antimigraine agent.
  • Chemical Name: 3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide succinate
  • Formula: C14H21N3O2S.C4H6O4
  • CAS Number: 103628-48-4
  • Solubility: Soluble to 100mM in water and to 75mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

Sumatriptan succinate
Sumatriptan succinate
413.49
>99