missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulphur Pieces, Puratronic™, 99.9995% (Metals basis), Alfa Aesar™

$109.28 - $1,372.55

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym acmc-20a47y, ad usum externum, lac, polymeric, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1034314
View Documents
Thermo Fisher Scientific
1034314
25g
Each for $109.28
 
AA1034322
View Documents
Thermo Fisher Scientific
1034322
100g
Each for $332.69
 
AA1034336
View Documents
Thermo Fisher Scientific
1034336
500g
Each for $1,372.55
 
Description

Description

Specifications

Chemical Identifiers

7704-34-9
32.06
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
sulfur
S
MFCD00085316
acmc-20a47y, ad usum externum, lac, polymeric, rubbervulcanizationagentis ot
CHEBI:26833
[S]

Specifications

7704-34-9
S
Pale yellow pieces
NINIDFKCEFEMDL-UHFFFAOYSA-N
sulfur
5362487
Pieces
Weak
25g
99.9995% (Metals basis)
MFCD00085316
acmc-20a47y, ad usum externum, lac, polymeric, rubbervulcanizationagentis ot
[S]
32.06
CHEBI:26833
248°C (478°F)
Yellow
Sulphur
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-722-6

RTECSNumber : WS4250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS