missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulphur Pieces, Puratronic™, 99.9995% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1034322

 View more versions of this product

Catalog No. AA1034322



Specifications

Specifications

Sulphur
99.9995% (Metals basis)
MFCD00085316
acmc-20a47y, ad usum externum, lac, polymeric, rubbervulcanizationagentis ot
[S]
32.06
CHEBI:26833
248°C (478°F)
Yellow
7704-34-9
S
Pale yellow pieces
NINIDFKCEFEMDL-UHFFFAOYSA-N
sulfur
5362487
Pieces
Weak
100g

Chemical Identifiers

7704-34-9
32.06
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
sulfur
S
MFCD00085316
acmc-20a47y, ad usum externum, lac, polymeric, rubbervulcanizationagentis ot
CHEBI:26833
[S]
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-722-6

RTECSNumber : WS4250000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents