Please login to your online account to display your discounted pricing

Sulfuric Acid RS, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
Name Note Reagent Solution
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 92.0
CAS 7664-93-9
CAS Min % 9.24
CAS Max % 9.62
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8154-1

 
ricca chemical company
RI81541
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8154-32

 
ricca chemical company
RI815432
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
CAS Min % 9.24
Chemical Name or Material Sulfuric Acid
Concentration or Composition Notes 10% (w/v)
Solvent or Matrix Aqueous Solution
Name Note Reagent Solution
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 92.0
CAS 7664-93-9
CAS Max % 9.62
Packaging Poly natural