Please login to your online account to display your discounted pricing

Sulfuric Acid, Multi-Compendial, N.F., J.T.Baker™

$168.71 - $887.38

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Purity Grade Multi-Compendial/NF
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004390

 
avantor performance
967103
Glass Bottle 2.5L Each for $887.38

02004391

 
avantor performance
967102
Glass Bottle 500mL Each for $168.71
Description & Specifications

Specifications

Purity Grade Multi-Compendial/NF
Formula Weight 98.08
Specific Gravity 1L = 1.84kg
Molecular Formula H2SO4
CAS 7664-93-9
Quantity 2.5L
Chemical Name or Material Sulfuric Acid
Packaging Glass Bottle