Please login to your online account to display your discounted pricing

Sulfuric acid, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sulfuric acid
CAS 7664-93-9
Physical Form Viscous Liquid
Molecular Formula H2SO4
Formula Weight 98.079
Synonym Oil of vitriol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14650252

 
avantor performance
287605
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK287618

 
avantor performance
287618
2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid
Formula Weight 98.079
Color Colorless to slightly yellow
Quantity 500mL
Density 1.84g/cm3
Synonym Oil of vitriol
Chemical Name or Material Sulfuric acid
Molecular Formula H2SO4
CAS 7664-93-9