Please login to your online account to display your discounted pricing

Sulfuric acid, Macron Fine Chemicals™


macron

Quantity 2.5L

Manufacturer: avantor performance  287618

 View more versions of this product

Catalog No. MK287618

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Sulfuric acid
Color Colorless to slightly yellow
Quantity 2.5L
CAS 7664-93-9
Physical Form Viscous Liquid
Molecular Formula H2SO4
Formula Weight 98.079
Synonym Oil of vitriol
Density 1.84g/cm3

Specifications

Chemical Name or Material Sulfuric acid
CAS 7664-93-9
Physical Form Viscous Liquid
Molecular Formula H2SO4
Formula Weight 98.079
Synonym Oil of vitriol