Please login to your online account to display your discounted pricing

Sulfuric Acid, For Phosphorus, Certified, 37.0% (v/v) ±1.0% , LabChem™

$36.77 - $36.77

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Max % 52.36
CAS 7664-93-9
CAS Max % 47.64
Name Note For Phosphorus
Purity Grade Certified Grade
Linear Formula H2SO4
Formula Weight 98.08
UN Number UN2796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC256201

 
lab chem
LC256201
500mL Each for $36.77
Description & Specifications

Specifications

Chemical Name or Material Sulfuric Acid
Color Clear, colorless
Quantity 500mL
Name Note For Phosphorus
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 52.36
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
CAS 7664-93-9
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 47.64
Purity Grade Certified Grade
Physical Form Liquid
Assay 37.0% ± 1.0% (v/v)
Linear Formula H2SO4
Formula Weight 98.08
UN Number UN2796
Density 1.37g/mL
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Specific Gravity 1.37g/mL
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.