Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfuric acid, Environmental Grade Plus, 93-98%

Chemical Identifiers

Chemical Name or Material Sulfuric acid
CAS 7664-93-9
Purity Grade Environmental Grade Plus
Formula Weight 98.08
MDL Number MFCD00064589
UN Number UN1830
Merck Index 14,8974
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38752AK

  SDS
alfa aesar™
38752AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38752AP

  SDS
alfa aesar™
38752AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid, Double Sub-Boiling Quartz Distillation
Formula Weight 98.08
Chemical Name or Material Sulfuric acid
TSCA TSCA
Quantity 250mL
Purity Grade Environmental Grade Plus
Assay 93 to 98%
MDL Number MFCD00064589
CAS 7664-93-9
UN Number UN1830
Merck Index 14,8974
Density 1.84g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SULFURIC ACID
Odor Acrid
RTECS Number WS5600000
Color Colorless to pale yellow
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.