Please login to your online account to display your discounted pricing

Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+% atom D, ACROS Organics™

$32.86 - $184.54

Chemical Identifiers

Chemical Name or Material Sulfuric acid-d2
Name Note 99.5+ atom % D
CAS 13813-19-9
Purity Grade For NMR
Assay 97.5 to 102.5%
Molecular Formula D2O4S
Linear Formula D2SO4
Formula Weight 100.09
MDL Number MFCD00003542
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175480050

  SDS
thermo fisher scientific
AC175480050
Glass bottle 5mL Each for $32.86

AC175480500

  SDS
thermo fisher scientific
175480500
Glass bottle 50mL Each for $184.54
Description & Specifications

Specifications

Chemical Name or Material Sulfuric acid-d2
Name Note 99.5+ atom % D
CAS 13813-19-9
Deuterium (D) 99.50% min.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
EINECS Number 237-464-0
TSCA TSCA
Density 1.8700g/mL
Purity Grade For NMR
Physical Form Liquid
Assay 97.5 to 102.5%
Quantity 5mL
Color Clear colorless
Molecular Formula D2O4S
Linear Formula D2SO4
Formula Weight 100.09
MDL Number MFCD00003542

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.