Please login to your online account to display your discounted pricing

Sulfuric acid-D2 96% sol. in D2O, deuteration deg. min. 99.5% for NMR, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 50mL
Packaging Glass bottle

Manufacturer: emd millipore corporation  8150600050

 View more versions of this product

Catalog No. M8150600050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 100.09
Quantity 50mL
Color Colorless
Physical Form Liquid
Name Note 96% sol. in D2O
Odor Odorless
Chemical Name or Material Sulfuric acid-D2 96% sol. in D2O
Vapor Pressure 0.001hPa (20°C)
Assay Percent Range 99.5% min.
Molecular Formula D2SO4
CAS 13813-19-9
UN Number 1830
Packaging Glass bottle

Specifications

Chemical Name or Material Sulfuric acid-D2 96% sol. in D2O
Name Note 96% sol. in D2O
CAS 13813-19-9
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula D2SO4
Formula Weight 100.09