Please login to your online account to display your discounted pricing

Sulfuric acid-D2 96% sol. in D2O, deuteration deg. min. 99.5% for NMR, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 25mL
Packaging Glass bottle

Manufacturer: emd millipore corporation  8150600025

 View more versions of this product

Catalog No. M8150600025

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sulfuric acid-D2 96% sol. in D2O
Name Note 96% sol. in D2O
CAS 13813-19-9
Odor Odorless
Packaging Glass bottle
Vapor Pressure 0.001hPa (20°C)
Physical Form Liquid
Quantity 25mL
Assay Percent Range 99.5% min.
Color Colorless
Molecular Formula D2SO4
Formula Weight 100.09
UN Number 1830

Specifications

Chemical Name or Material Sulfuric acid-D2 96% sol. in D2O
Name Note 96% sol. in D2O
CAS 13813-19-9
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula D2SO4
Formula Weight 100.09