Please login to your online account to display your discounted pricing

Sulfuric acid-D2 96% sol. in D2O, deuteration deg. min. 99.5% for NMR, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$228.03 - $380.09

Chemical Identifiers

Chemical Name or Material Sulfuric acid-D2 96% sol. in D2O
Name Note 96% sol. in D2O
CAS 13813-19-9
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula D2SO4
Formula Weight 100.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M8150600050

 
emd millipore corporation
8150600050
Glass bottle 50mL Each for $380.09

M8150600025

 
emd millipore corporation
8150600025
Glass bottle 25mL Each for $228.03
Description & Specifications

Specifications

Chemical Name or Material Sulfuric acid-D2 96% sol. in D2O
Name Note 96% sol. in D2O
CAS 13813-19-9
Odor Odorless
Packaging Glass bottle
Vapor Pressure 0.001hPa (20°C)
Physical Form Liquid
Quantity 50mL
Assay Percent Range 99.5% min.
Color Colorless
Molecular Formula D2SO4
Formula Weight 100.09
UN Number 1830