Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 72% (w/w), 24.0N ±0.1N (12M), LabChem™

$93.80 - $93.80

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
CAS Max % 72
CAS 7732-18-5
CAS Max % 28
Purity Grade Certified Grade
Linear Formula H2SO4
Formula Weight 98.08
UN Number UN1830
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC256452

 
lab chem
LC256452
1L Each for $93.80
Description & Specifications

Specifications

Formula Weight 98.08
Purity Grade Certified Grade
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Sulfuric Acid
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
UN Number UN1830
Specific Gravity 1.634g/mL
CAS Max % 72
Identification Passes Test
Traceability to NIST Traceable to NIST
Packaging Amber Glass
Solubility Soluble in water
Physical Form Liquid
Color Clear, colorless
CAS 7664-93-9
Quantity 1L
Storage Information Room Temperature
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Health Hazard 1 Danger
Assay 72% (w/w)
Linear Formula H2SO4
CAS Max % 28
Density 1.634g/mL
Concentration 24.0N ±0.1N (12M)
CAS 7732-18-5
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.