missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 25.0% (v/v) ±0.5%, LabChem™

Manufacturer:  Lab Chem LC256103

 View more versions of this product

Catalog No. LC256103



Specifications

Specifications

Sulfuric Acid
2.5L
64.95,35.05
H2SO4
acide sulfurique
OS(=O)(=O)O
1.26g/mL
1118
98.08
25.0% ± 0.5% (v/v)
Passes Test
Soluble in water
Room Temperature
Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Colorless
7664-93-9,7732-18-5
H2O4S
UN2796
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
98.072
CHEBI:26836
Liquid
Certified
Clear Glass
1.26g/mL
Traceable to NIST

Chemical Identifiers

7664-93-9
98.072
acide sulfurique
CHEBI:26836
OS(=O)(=O)O
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Documents