Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 10.0N ±0.1N (5.0M), LabChem™

$63.77 - $141.46

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Max % 61.89
CAS 7664-93-9
CAS Max % 38.11
Purity Grade Certified Grade
Linear Formula H2SO4
Formula Weight 98.08
UN Number UN2796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC258702

 
lab chem
LC258702
1L Each for $63.77

LC258703

 
lab chem
LC258703
2.5L Each for $141.46
Description & Specifications

Specifications

Formula Weight 98.08
Purity Grade Certified Grade
UN Number UN2796
Solubility Soluble in water
Health Hazard 1 Danger
Concentration 10.0N ±0.1N (5.0M)
Identification Passes Test
Quantity 1L
Density 1.29g/mL
Color Clear, colorless
Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
CAS Max % 61.89
Traceability to NIST Traceable to NIST
Storage Information Room Temperature
Physical Form Liquid
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Linear Formula H2SO4
Specific Gravity 1.29g/mL
Decomposition Information Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Packaging Poly Bottle
CAS Max % 38.11
CAS 7732-18-5
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.