missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 10.0% (w/w) ±0.5%, LabChem™

$38.34 - $1,901.09

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.072
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym acide sulfurique
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(=O)(=O)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC255902 View Documents Lab Chem
LC255902
1L Each for $38.34
 
LC255903 View Documents Lab Chem
LC255903
2.5L Each for $84.97
 
LC255906 View Documents Lab Chem
LC255906
10L Each for $169.74
 
LC255905 View Documents Lab Chem
LC255905
20L Each for $303.80
 
LC25590200 View Documents Lab Chem
LC25590200
200L Each for $1,901.09
 
Specifications

Chemical Identifiers

7664-93-9
98.072
acide sulfurique
CHEBI:26836
OS(=O)(=O)O
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid

Specifications

Colorless
7664-93-9,7732-18-5
H2O4S
UN2796
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
98.072
CHEBI:26836
Liquid
Certified
Poly Bottle
1.07g/mL
Traceable to NIST
Sulfuric Acid
1L
90,10
H2SO4
acide sulfurique
OS(=O)(=O)O
1.07g/mL
1118
98.08
10.0% ± 0.5% (w/w)
Passes Test
Soluble in water
Room Temperature
Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger