missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Certified, 10.0% (v/v) ±0.5%, LabChem™
$53.00 - $126.10
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| Colorless | |
| 1.11g/mL | |
| Passes Test | |
| 1.11g/mL | |
| Sulfur compounds; Thermal decomposition generates: Corrosive vapors | |
| H2O4S | |
| H2SO4 | |
| UN2796 | |
| Soluble in water | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| 10.0% ±0.5% (v/v) | |
| Sulfuric Acid |
| 7664-93-9,7732-18-5 | |
| 84.07,15.93 | |
| Poly Bottle | |
| Traceable to NIST | |
| 500 mL | |
| Liquid | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08 | |
| Certified |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto
Danger
Recommended Storage : Room Temperature