missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 1.000N ±0.005N (0.5M), LabChem™

$19.29 - $187.00

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.072
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym acide sulfurique
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(=O)(=O)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC257701 View Documents Lab Chem
LC257701
500mL Each for $19.29
 
LC257702 SDS View Documents Lab Chem
LC257702
1L Each for $27.83
 
LC257704 View Documents Lab Chem
LC257704
4L Each for $65.20
 
LC257706 View Documents Lab Chem
LC257706
10L Each for $124.71
 
LC257705 View Documents Lab Chem
LC257705
20L Each for $187.00
 
Specifications

Chemical Identifiers

7664-93-9
98.072
acide sulfurique
CHEBI:26836
OS(=O)(=O)O
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid

Specifications

Colorless
7664-93-9,7732-18-5
H2O4S
UN2796
QAOWNCQODCNURD-UHFFFAOYSA-N
1.000N ± 0.005N
1.03g/mL
1118
98.08
Certified
Poly Bottle
1.03g/mL
Traceable to NIST
Sulfuric Acid, 1.0N (0.5M)
500mL
95.24,4.76
H2SO4
acide sulfurique
OS(=O)(=O)O
sulfuric acid
98.072
CHEBI:26836
Liquid
Passes Test
Soluble in water
Room Temperature
Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger