Learn More
Sulfuric Acid, Certified, 1.000N ±0.005N (0.5M), LabChem™
Supplier: LabChem LC257705
Specifications
Sulfuric Acid | |
7664-93-9,7732-18-5 | |
1.03g/mL | |
Passes Test | |
1.03g/mL | |
Sulfur compounds; Thermal decomposition generates: Corrosive vapors | |
H2O4S | |
H2SO4 | |
UN2796 | |
Soluble in water | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Certified |
Colorless | |
1.000N ±0.005N (0.5M) | |
95.24,4.76 | |
Cubitainer™ | |
Traceable to NIST | |
20 L | |
Liquid | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature