Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 0.1250N ±0.0010N (0.0625M), LabChem™

$91.31 - $91.31

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Max % 99.39
CAS 7664-93-9
CAS Max % 0.61
Purity Grade Certified Grade
Linear Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC256852

 
lab chem
LC256852
1L Each for $91.31
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 98.08
Chemical Name or Material Sulfuric Acid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Sulfur compounds
Physical Form Liquid
Linear Formula H2SO4
CAS 7732-18-5
Concentration 0.1250N ±0.0010N (0.0625M)
Density 1g/mL
CAS Max % 99.39
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7664-93-9
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Identification Passes Test
CAS Max % 0.61
Packaging Poly Bottle
Viscosity 1.01cSt
Color Clear, colorless
Quantity 1L
Safety and Handling

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Solution is not hazardous.