Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 0.1000N ±0.0005N (0.05M), LabChem™

$23.63 - $180.00

Chemical Identifiers

Chemical Name or Material Sulfuric Acid, 0.1N (0.05M)
CAS 7732-18-5
CAS Max % 99.51
CAS 7664-93-9
CAS Max % 0.49
Purity Grade Certified Grade
Linear Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC256802

 
lab chem
LC256802
1L Each for $27.00

LC256801

 
lab chem
LC256801
500mL Each for $23.63

LC256804

 
lab chem
LC256804
4L Each for $55.31

LC256805

 
lab chem
LC256805
20L Each for $180.00

LC256806

 
lab chem
LC256806
10L Each for $116.97
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 98.08
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Chemical Name or Material Sulfuric Acid, 0.1N (0.05M)
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Sulfur compounds
Physical Form Liquid
Linear Formula H2SO4
CAS 7732-18-5
Concentration 0.1000N ± 0.0005N
Density 1g/mL
CAS Max % 99.51
CAS 7664-93-9
Identification Passes Test
CAS Max % 0.49
Packaging Poly Bottle
Viscosity 1.01cSt
Color clear, colorless solution
Quantity 1L
Safety and Handling

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at

GHS H Statement
Solution is not hazardous.