missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, Certified, 0.1000N ±0.0005N (0.05M), LabChem™
Supplier: LabChem LC256806
Specifications
Sulfuric Acid | |
7664-93-9,7732-18-5 | |
1g/mL | |
Passes Test | |
1g/mL | |
Sulfur compounds | |
1.01cSt | |
Liquid | |
MFCD00064589 | |
Soluble in water | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Certified |
Colorless | |
0.1000N ±0.0005N (0.05M) | |
99.51,0.49 | |
Cubitainer™ | |
Traceable to NIST | |
10 L | |
H2O4S | |
H2SO4 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage : Room Temperature