missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 0.0200N ±0.0002N (0.01M), LabChem™

Manufacturer:  Lab Chem LC256601

 View more versions of this product

Catalog No. LC256601



Specifications

Specifications

Sulfuric Acid
Colorless
7664-93-9,7732-18-5
H2O4S
acide sulfurique
OS(=O)(=O)O
sulfuric acid
98.072
CHEBI:26836
Liquid
Passes Test
Soluble in water
Room Temperature
Sulfur compounds
1cSt
500mL
99.9,0.1
H2SO4
QAOWNCQODCNURD-UHFFFAOYSA-N
0.0200N ±0.0002N (0.01M)
1g/mL
1118
98.08
Certified
Poly Bottle
1g/mL
Traceable to NIST

Chemical Identifiers

7664-93-9
98.072
acide sulfurique
CHEBI:26836
OS(=O)(=O)O
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Documents