missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 0.0100N ±0.0001N (0.005M), LabChem™

Supplier:  LabChem LC256552

 View more versions of this product

Catalog No. LC256552



Description

Description

Specifications

Specifications

Sulfuric Acid
7664-93-9,7732-18-5
1g/mL
Passes Test
1g/mL
Sulfur compounds
1cSt
Liquid
MFCD00064589
Soluble in water
OS(O)(=O)=O
98.07
CHEBI:26836
Certified
Colorless
0.0100N ±0.0001N (0.005M)
99.95,0.05
Poly Bottle
Traceable to NIST
1 L
H2O4S
H2SO4
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions