Please login to your online account to display your discounted pricing

Sulfuric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

$152.52 - $237.13

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004373

 
avantor performance
939005
Poly Bottle 2.5L Each for $237.13

02004380

 
avantor performance
939001
Poly Bottle 500mL Each for $152.52
Description & Specifications

Specifications

Formula Weight 98.08
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material Sulfuric Acid
Specific Gravity 1L = 1.84kg
Molecular Formula H2SO4
CAS 7664-93-9
Packaging Poly Bottle
Quantity 2.5L
Purity Grade BAKER INSTRA-ANALYZED™ Plus

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; NOTE: Intended for nonfuel end purposes only