Please login to your online account to display your discounted pricing

Sulfuric acid, for analysis ACS, 95% solution in water, ACROS Organics™


Quantity 2.5L
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC424520026

 View more versions of this product

Catalog No. AC424520026

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sulfuric acid
Color Clear
Quantity 2.5L
Ammonium 2ppm max.
CAS 7664-93-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Arsenic (As) 0.01ppm max.
Purity Grade ACS Reagent Grade
Physical Form Liquid Free From Suspended or Insoluble Matter
Assay 95.0 to 98.0%
Assay Percent Range 95%
Molecular Formula H2O4S
Linear Formula H2SO4
Formula Weight 98.07
Chlorine (Cl) 0.2ppm max.
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Density 1.8300g/mL
Heavy Metals (as Pb) 1ppm max.
Iron (Fe) 0.2ppm max.
Mercury (Hg) 5ppb max.
Nitrate 0.5ppm max.
Packaging Plastic bottle
Residue after Ignition 0.0005% max.
Substances Reducing Permanganate 2ppm max. (as SO2)

Specifications

Chemical Name or Material Sulfuric acid
CAS 7664-93-9
Purity Grade ACS Reagent Grade
Assay 95.0 to 98.0%
Assay Percent Range 95%
Molecular Formula H2O4S
Linear Formula H2SO4
Formula Weight 98.07
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents