Please login to your online account to display your discounted pricing

Sulfuric Acid, ACS Reagent Grade, Ricca Chemical


Quantity 500mL
Packaging Glass amber

Manufacturer: ricca chemical company  RIRABS0030500C

 View more versions of this product

Catalog No. RABS0030500

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Liquid
Color Clear colorless
Quantity 500mL
CAS 7664-93-9
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass amber
Chemical Name or Material Sulfuric Acid

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Liquid