Please login to your online account to display your discounted pricing

Sulfuric Acid ACS, EMD Millipore


Quantity 2.5L
Packaging Poly Coated Glass Bottle

Manufacturer: emd millipore corporation  SX1244-7

 View more versions of this product

Catalog No. MSX12447

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Density 1.84g/cm3 (20°C)
Packaging Poly Coated Glass Bottle
Vapor Pressure 0.0001 hPa (20°C)
Quantity 2.5L
Chemical Name or Material Sulfuric Acid
Name Note Suprapur™
CAS 7664-93-9
Purity Grade ACS
Assay Percent Range 96%
Molecular Formula H2O4S
Linear Formula H2SO4
Formula Weight 98.8g/mol
Recommended Storage Store without restrictions

Specifications

Chemical Name or Material Sulfuric Acid
Name Note Suprapur™
CAS 7664-93-9
Purity Grade ACS
Assay Percent Range 96%
Molecular Formula H2O4S
Linear Formula H2SO4
Formula Weight 98.8g/mol
Accessories

Store without restrictions