missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid ACS, 95 to 98%, MilliporeSigma™
Supplier: Technidata SX12446
Specifications
Sulfuric Acid | |
-20°C | |
335°C | |
Glass Bottle | |
500 mL | |
H2SO4 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
95 to 98% |
Suprapur™ | |
7664-93-9 | |
1.84 g/cm3 (20°C) | |
0.0001 hPa (20°C) | |
H2O4S | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
ACS |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
EINECSNumber : 231-639-5
RTECSNumber : WS5600000
Recommended Storage : Store without restrictions