Please login to your online account to display your discounted pricing

Sulfuric Acid ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Purity Grade ACS Grade
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

14650380

 
avantor performance
287646
Glass Container 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK5557212

 
avantor performance
555746
SAFEMOR 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK5557500

 
avantor performance
555714
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric Acid
Quantity 2.5L
CAS 7664-93-9
Purity Grade ACS Grade
Molecular Formula H2SO4
Formula Weight 98.08
Packaging Glass Container