Please login to your online account to display your discounted pricing

Sulfuric Acid ACS AR, Macron Fine Chemicals™


macron

Quantity 2.5L
Packaging SAFEMOR

Manufacturer: avantor performance  555746

 View more versions of this product

Catalog No. MK5557212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 98.08
Quantity 2.5L
Chemical Name or Material Sulfuric Acid
Molecular Formula H2SO4
CAS 7664-93-9
Packaging SAFEMOR

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Purity Grade ACS Grade
Molecular Formula H2SO4
Formula Weight 98.08