Please login to your online account to display your discounted pricing

Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sulfuric acid
CAS 7664-93-9
Physical Form Liquid
Molecular Formula H2SO4
Formula Weight 98.08
MDL Number MFCD00064589
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88568LQ

  SDS
alfa aesar
88568LQ
1 unit CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA88568LS

  SDS
alfa aesar
88568LS
6 units CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.08
Quantity 1 unit
Color Colorless to pale yellow
DOT Information DOT Class: 8, Packaging Group: II
CAS 7664-93-9
EINECS Number 231-639-5
Physical Form Liquid
Chemical Name or Material Sulfuric acid
Molecular Formula H2SO4
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Health Hazard 1 H314: Causes severe skin burns and eye damage.
Appearance Colorless to pale yellow liquid
MDL Number MFCD00064589
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Odor Acrid
Concentration or Composition (by Analyte or Components) Sulfuric acid: 55%; Water: 45%
Safety and Handling

H314: Causes severe skin burns and eye damage.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).