missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfuric acid, 99.9999% (metals basis), 92% min

Manufacturer:  Alfa Aesar™ 11000AP

 View more versions of this product

Catalog No. AA11000AP



Specifications

Specifications

Sulfuric acid
500mL
(metals basis)
MFCD00064589
14,8974
QAOWNCQODCNURD-UHFFFAOYSA-N
≤92%
1.84g/mL
1118
98.08
99.9999%
Colorless to Yellow
7664-93-9
H2O4S
UN1830
acide sulfurique
OS(=O)(=O)O
sulfuric acid
98.072
CHEBI:26836
Liquid
Acrid

Chemical Identifiers

7664-93-9
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
acide sulfurique
CHEBI:26836
OS(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SULFURIC ACID

RTECSNumber : WS5600000

TSCA : TSCA

Documents