missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 133610026

 View more versions of this product

Catalog No. AC133610026



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sulfuric acid
96% Solution in Water
7664-93-9
1.8300g/mL
1.83
2.5 L
H2O4S
H2SO4
01,470; 05,633; 06,558; 07,347; 09,441
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
Extra Pure
Colorless
10.0°C
290.0°C
Plastic Bottle
96%
21 mPa.s (25°C)
Oily Liquid
MFCD00064589
15, 9104
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
OS(O)(=O)=O
98.07
CHEBI:26836
96%

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only