Please login to your online account to display your discounted pricing

Sulfuric Acid, 95.0 to 98.0%, N.F. - F.C.C., J.T.Baker™

$130.46 - $835.02

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Purity Grade FCC/NF
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004387

 
avantor performance
967503
Glass Bottle 2.5L Each for $835.02

02004388

 
avantor performance
967502
Glass Bottle 500mL Each for $130.46
Description & Specifications

Specifications

Purity Grade FCC/NF
Assay 95.0 to 98.0%
Formula Weight 98.08
Chemical Name or Material Sulfuric Acid
Specific Gravity 1L = 1.84kg
Molecular Formula H2SO4
CAS 7664-93-9
Packaging Glass Bottle
Quantity 2.5L