Please login to your online account to display your discounted pricing

Sulfuric acid, 93-98%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sulfuric acid
Name Note Environmental Grade
CAS 7664-93-9
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range 93 - 98%
Molecular Formula H2SO4
Formula Weight 98.08
MDL Number MFCD00064589
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38751AP

  SDS
alfa aesar
38751AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38751K4

  SDS
alfa aesar
38751K4
2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric acid
Color Colorless to pale yellow
Quantity 500mL
Name Note Environmental Grade
CAS 7664-93-9
Health Hazard 1 H314: Causes severe skin burns and eye damage.
Appearance Colorless to pale yellow liquid
DOT Information DOT Class: 8, Packaging Group: II
EINECS Number 231-639-5
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range 93 - 98%
Molecular Formula H2SO4
Formula Weight 98.08
MDL Number MFCD00064589
Odor Acrid
Safety and Handling

H314: Causes severe skin burns and eye damage.