missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, 90% (w/w), Ricca Chemical
Supplier: Ricca Chemical Company R8194000500A
Specifications
Sulfuric Acid | |
94.0,5.0 | |
Natural Poly Bottle | |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
90% (w/w) Aqueous Solution |
7664-93-9,7732-18-5 | |
95.0,6.0 | |
500 mL | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(=O)(=O)O | |
98.072 | |
CHEBI:26836 |
Safety and Handling
ShelfLife : 24 Months