Please login to your online account to display your discounted pricing

Sulfuric Acid, 90% (w/w) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR819400020F

 View more versions of this product

Catalog No. R819400020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 20L
CAS 7664-93-9
CAS Max % 95.0
CAS Min % 5.0
CAS Max % 6.0
CAS Min % 94.0
Shelf Life 24 Months
Concentration or Composition (by Analyte or Components) 90% (w/w) Aqueous Solution
Packaging Cubitainer
CAS 7732-18-5
Chemical Name or Material Sulfuric Acid

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
CAS Min % 94.0
CAS Max % 95.0
CAS 7732-18-5
CAS Min % 5.0
CAS Max % 6.0
Physical Form Liquid
Accessories

24 Months