missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric acid, ≥90% solution, J.T.Baker™

Manufacturer:  Avantor Performance H37809

 View more versions of this product

Catalog No. MKH37820

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sulfuric acid
20L
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
OS(=O)(=O)O
98.072
CHEBI:26836
Viscous Liquid
Colorless to Yellow
7664-93-9
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1.84g/cm3
sulfuric acid
1118
98.079

Chemical Identifiers

7664-93-9
98.072
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(=O)(=O)O
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
Documents