missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, 80% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company R81900004A
Specifications
Sulfuric Acid | |
7664-93-9,7732-18-5 | |
Natural Poly Bottle | |
80.0 ± 0.5% (v/v) | |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
APHA/EPA Certified Reagent |
80% (v/v) Aqueous Solution (4 + 1) | |
Odorless | |
Aqueous Solution | |
4 L | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(=O)(=O)O | |
98.072 | |
CHEBI:26836 |
Chemical Identifiers
7664-93-9 | |
98.072 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(=O)(=O)O |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |